MilliporeSigma
  • In vitro effect of H 2 O 2, some transition metals and hydroxyl radical produced via fenton and fenton-like reactions, on the catalytic activity of AChE and the Hydrolysis of ACh
In vitro effect of H 2 O 2, some transition metals and hydroxyl radical produced via fenton and fenton-like reactions, on the catalytic activity of AChE and the Hydrolysis of ACh
Neurochemical Research (2014)
Mendez-Garrido A, et al