MilliporeSigma
  • Interspecies Interactions Determine the Impact of the Gut Microbiota on Nutrient Allocation in Drosophila melanogaster
Interspecies Interactions Determine the Impact of the Gut Microbiota on Nutrient Allocation in Drosophila melanogaster
Applied and Environmental Microbiology (2014)
Newell PD, et al.