MilliporeSigma
  • Development of a fast and sensitive method for measuring multiple neonicotinoid insecticide residues in soil and the application in parks and residential areas.
Development of a fast and sensitive method for measuring multiple neonicotinoid insecticide residues in soil and the application in parks and residential areas.
Analytica Chimica Acta (2018)
Zhou Y, et al.