MilliporeSigma
  • Highly Fluorescent Chiral N?S?Doped Carbon Dots from Cysteine: Affecting Cellular Energy Metabolism.
Highly Fluorescent Chiral N?S?Doped Carbon Dots from Cysteine: Affecting Cellular Energy Metabolism.
Angewandte Chemie (International Edition in English) (2018)
Li F, et al.