MilliporeSigma
  • PbS-NiO nanocomposite material with enhanced magnetic, photocatalytic and antifungal properties
PbS-NiO nanocomposite material with enhanced magnetic, photocatalytic and antifungal properties
Materials Science and Engineering, B (2018)
Suganya M, et al.