MilliporeSigma
  • Hierarchical NiO hollow microspheres assembled from nanosheet-stacked nanoparticles and their application in a gas sensor
Hierarchical NiO hollow microspheres assembled from nanosheet-stacked nanoparticles and their application in a gas sensor
Royal Society of Chemistry Advances (2012)
Zhu G, et al.