MilliporeSigma
  • Differential spectrophotometric determination of tyrosine and tryptophan in pharmaceutical amino acid solutions
Differential spectrophotometric determination of tyrosine and tryptophan in pharmaceutical amino acid solutions
Journal of Pharmaceutical Sciences (1978)
Demeester J, et al.