MilliporeSigma
  • Electrical and optical properties of Al doped cadmium oxide thin films deposited by radio frequency magnetron sputtering
Electrical and optical properties of Al doped cadmium oxide thin films deposited by radio frequency magnetron sputtering
Solar Energy Materials and Solar Cells (2007)
Saha B, et al.