MilliporeSigma
  • Method development and validation for the simultaneous estimation of lycopene and ubidecarenone by RP-HPLC in combined pharmaceutical dosage form
Method development and validation for the simultaneous estimation of lycopene and ubidecarenone by RP-HPLC in combined pharmaceutical dosage form
Journal of Drug Delivery and Therapeutics (2016)
SVS, et al.