MilliporeSigma
  • Denaturant-Dependent Conformational Changes in a beta-Trefoil Protein: Global and Residue-Specific Aspects of an Equilibrium Denaturation Process
Denaturant-Dependent Conformational Changes in a beta-Trefoil Protein: Global and Residue-Specific Aspects of an Equilibrium Denaturation Process
Biochemistry (2009)
Latypov RF, et al.