MilliporeSigma
  • In-toilet disinfection of fresh fecal sludge with ammonia naturally present in excreta
In-toilet disinfection of fresh fecal sludge with ammonia naturally present in excreta
Journal of Water, Sanitation and Hygiene for Development (2016)
Ogunyoku TA, et al.