MilliporeSigma
  • The in vitro effect of gefitinib (Iressa) alone and in combination with cytotoxic chemotherapy on human solid tumours
The in vitro effect of gefitinib (Iressa) alone and in combination with cytotoxic chemotherapy on human solid tumours
BMC Cancer (2004)
Knight LA, et al.