MilliporeSigma
  • Protein conformation changes on block copolymer surfaces detected by antibody-functionalized atomic force microscope tips.
Protein conformation changes on block copolymer surfaces detected by antibody-functionalized atomic force microscope tips.
Journal of biomedical materials research. Part A (2011-10-06)
Manuel L B Palacio, Scott R Schricker, Bharat Bhushan
ABSTRACT
Conformational changes of fibronectin (Fn) deposited on poly(methyl methacrylate) and poly(acrylic acid) block copolymers with identical chemical compositions were detected using an antibody-functionalized atomic force microscope (AFM) tip. Based on the antibody-protein adhesive force maps and phase imaging, it was found that the nanomorphology of the triblock copolymer is conducive to the exposure of the arginine-glycine-aspartic acid (RGD) groups in Fn. For the first time, X-ray photoelectron spectroscopy was used to elucidate surface chemical composition and confirm AFM results. The findings demonstrate that block copolymer nanomorphology can be used to regulate protein conformation and potentially cellular response.
Sigma-Aldrich
Anti-Fibronectin Frag Antibody, cell attachment fragment, clone 3E3, clone 3E3, Chemicon®, from mouse