MilliporeSigma
  • The concentration of tumor necrosis factor-α determines its protective or damaging effect on liver injury by regulating Yap activity.
The concentration of tumor necrosis factor-α determines its protective or damaging effect on liver injury by regulating Yap activity.
Cell death & disease (2020-01-29)
Shanmin Zhao, Jinghua Jiang, Yingying Jing, Wenting Liu, Xue Yang, Xiaojuan Hou, Lu Gao, Lixin Wei
ABSTRACT
Previous studies have shown that tumor necrosis factor (TNF)-α is a mediator of hepatotoxicity in liver injury. Moreover, TNF-α has also been reported to have a protective effect in liver regeneration, yet the function of TNF-α during liver injury remains controversial. Here, we report that the concentration of TNF-α determines its functions. High concentrations of TNF-α could aggravate LPS-induced liver injury. However, the TNF-α level was unchanged during APAP-induced liver injury, which exerted a protective effect. We expected that the concentration of TNF-α may affect its function. To test this hypothesis, TNF-α-/- rats or hepatocyte cells were treated with different concentrations of TNF-α. We found low TNF-α could reduce the levels of ALT and AST in the plasma of TNF-α-/- rats and promote the proliferation of hepatocyte cells. However, the levels of ALT and AST increased gradually with increasing TNF-α concentration after reaching the lowest value. Moreover, we showed that TNF-α affects the cell proliferation and cell death of hepatocytes by regulating Yap activity. Low TNF-α promoted Yap1 nuclear translocation, triggering the proliferation of hepatocytes. However, high TNF-α triggered the phosphorylation and inactivation of Yap1, preventing its nuclear import and consequently promoting cell death. Collectively, our findings provide novel evidence that the concentration of TNF-α is an important factor affecting its function in liver injury, which may provide a reference for the clinical treatment of liver injury.
Sigma-Aldrich
Anti-phospho-Mst1/2 (pThr183) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-LATS1 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-LATS1/2 (pThr1079/1041) antibody produced in rabbit, affinity isolated antibody