MilliporeSigma
  • Control of catenation in CuTATB-n metal-organic frameworks by sonochemical synthesis and its effect on CO 2 adsorption
Control of catenation in CuTATB-n metal-organic frameworks by sonochemical synthesis and its effect on CO 2 adsorption
Journal of Materials Chemistry (2011)
Kim J, et al.