MilliporeSigma
  • Synthesis and Basicity Studies of Quinolino[7,8-h]quinoline Derivatives.
Synthesis and Basicity Studies of Quinolino[7,8-h]quinoline Derivatives.
The Journal of organic chemistry (2020-08-14)
Gareth J Rowlands, Rebecca J Severinsen, Jenna K Buchanan, Karl J Shaffer, Heather T Jameson, Nishani Thennakoon, Ivo Leito, Märt Lõkov, Agnes Kütt, Robert Vianello, Ines Despotović, Nena Radić, Paul G Plieger
ABSTRACT
Quinolino[7,8-h]quinoline is a superbasic compound, with a pKaH in acetonitrile greater than that of 1,8-bis(dimethylaminonaphthalene) (DMAN), although its synthesis and the synthesis of its derivatives can be problematic. The use of halogen derivatives 4,9-dichloroquinolino[7,8-h]quinoline (16) and 4,9-dibromoquinolino[7,8-h]quinoline (17) as precursors has granted the formation of a range of substituted quinolinoquinolines. The basicity and other properties of quinolinoquinolines can be modified by the inclusion of suitable functionalities. The experimentally obtained pKaH values of quinolino[7,8-h]quinoline derivatives show that N4,N4,N9,N9-tetraethylquinolino[7,8-h]quinoline-4,9-diamine (26) is more superbasic than quinolino[7,8-h]quinoline. Computationally derived pKaH values of quinolinoquinolines functionalized with dimethylamino (NMe2), 1,1,3,3-tetramethylguanidino (N═C(NMe2)2) or N,N,N',N',N″,N″-hexamethylphosphorimidic triamido (N═P(NMe2)3) groups are significantly greater than those of quinolino[7,8-h]quinoline. Overall, electron-donating functionalities are observed to increase the basicity of the quinolinoquinoline moiety, while the substitution of electron-withdrawing groups lowers the basicity.
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%