MilliporeSigma
  • Elevated air temperature damage to photosynthetic apparatus alleviated by enhanced cyclic electron flow around photosystem I in tobacco leaves.
Elevated air temperature damage to photosynthetic apparatus alleviated by enhanced cyclic electron flow around photosystem I in tobacco leaves.
Ecotoxicology and environmental safety (2020-08-18)
Che Yanhui, Wang Hongrui, Zhang Beining, Gao Shixing, Wang Zihan, Wang Yue, Zhang Huihui, Sun Guangyu
ABSTRACT
High temperature can lead to increased production of excess light energy, thus reducing photosynthetic capacity in plants. Photosynthetic cyclic electron flow (CEF) in photosystem I (PSI) can effectively protect photosystems, but its physiological mechanism under high temperature is poorly understood. In this study, antimycin A (AA) and thenoyltrifluoroacetone (TTFA) were used to inhibit PGR5-and NDH-dependent CEF pathways, respectively, to reveal the photoprotective functions of CEF for PSII in tobacco leaves under high temperature stress (37 °C, HT). High temperatures caused decreases in maximal photochemistry efficiency (Fv/Fm) and damaged photosystem II (PSII) in tobacco leaves. Under AA inhibition of PGR5-dependent CEF, high temperature increased the fluorescence intensity of point O (Fo) in OJIP curves, i.e., the energy absorption per active reaction center (ABS/RC), the trapping rate of the reaction center (TRo/RC), and the electron transport efficiency per reaction center (ETo/RC) in tobacco leaves. High temperature induced an increase in the hydrogen peroxide content and a decrease in pigment content in tobacco leaves. Under the high temperature treatment, inhibition of PGR5-dependent CEF reduced the activities of the PSII reaction center significantly, destroyed the oxygen-evolving complex (OEC), and impeded photosynthetic electron transfer from PSII to the plastoquinone (PQ) pool in tobacco leaves. The TTFA treatment inhibited the NDH-dependent pathway under high temperature conditions, with the relative fluorescence intensity of point I (VI) decreased significantly, and the content of hydrogen peroxide and superoxide anion increased significantly. Additionally, Fo and the redox degree of the PSII donor side (Wk) increased, and pigment content decreased compared to the control, but with little change compared to high temperature treatment, indicating that the inhibition of the NDH-dependent pathway directly weakened the capacity of the PQ pool to lead to the accumulation of reactive oxygen species (ROS) in tobacco leaves. In conclusion, CEF alleviated damage to the photosynthetic apparatus in tobacco leaves by increasing PSII heat dissipation, reducing ROS production, and maintaining the stability of the PQ pool to accommodate photosynthetic electron flow.
Product Description
Supelco
Chlorophyll b, analytical standard