MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Involvement of tryptase-positive mast cells and angiogenesis in the growth of inflammatory odontogenic cysts.
Involvement of tryptase-positive mast cells and angiogenesis in the growth of inflammatory odontogenic cysts.
Brazilian oral research (2021-06-03)
Hugo Costa Neto, Ana Luiza Dias Leite de Andrade, Andréia Ferreira do Carmo, Roseana de Almeida Freitas, Hébel Cavalcanti Galvão
ABSTRACT
Inflammatory periapical lesions are characterized by infiltration of different immune cell types, the functions of which depend on an effective vascular network. This study aimed to evaluate the mast cells density (MCD) in inflamatory odontogenic cysts capsules concerning microvascular density (MVD), microvascular area (MVA), and microvascular perimeter (MVP), and correlate such findings with the type of lesion, intensity of the inflammatory infiltrate, and thickness of the epithelial lining. Twenty inflamatory dentigerous cysts (IDCs), twenty radicular cysts (RCs), and twenty residual radicular cysts (RRCs) were submitted to immunohistochemical analysis using anti-tryptase and anti-CD34 antibodies. RCs exhibited the highest MCD, MVD, MVA, and MVP indexes (p = < 0.001, p = 0.008, p = 0.003 and p = < 0.001, respectively), and lesions with inflammatory infiltrate grade III showed the highest MVD (p = 0.044). Considering epithelial thickness, a higher MVP index was identified in lesions with hyperplastic epithelium (p = 0.018). In IDCs, RCs, and RRCs, a strong positive correlation was observed between MVA and MVP (r = 0.950 and p = < 0.001; r = 0.914 and p = < 0.001; r = 0.713 and p = < 0.001, respectively). In IDCs, a moderate correlation was observed between MCD and both MVA and MVP (r = 0.660 and p = 0.002; r = 0.634 and p = 0.003, respectively). These results suggest that tryptase-positive mast cells might play an important role in the angiogenic activity of IDCs, while RCs had the highest indexes. Our findings also confirmed that the intensity of the inflammatory infiltrate and epithelial thickness influence angiogenesis.
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride hydrate, ≥96%