MilliporeSigma
  • Multiomics of World Trade Center Particulate Matter-induced Persistent Airway Hyperreactivity. Role of Receptor for Advanced Glycation End Products.
Multiomics of World Trade Center Particulate Matter-induced Persistent Airway Hyperreactivity. Role of Receptor for Advanced Glycation End Products.
American journal of respiratory cell and molecular biology (2020-04-22)
Syed H Haider, Arul Veerappan, George Crowley, Erin J Caraher, Dean Ostrofsky, Mena Mikhail, Rachel Lam, Yuyan Wang, Maria Sunseri, Sophia Kwon, David J Prezant, Mengling Liu, Ann Marie Schmidt, Anna Nolan
ABSTRACT
Pulmonary disease after World Trade Center particulate matter (WTC-PM) exposure is associated with dyslipidemia and the receptor for advanced glycation end products (RAGE); however, the mechanisms are not well understood. We used a murine model and a multiomics assessment to understand the role of RAGE in the pulmonary long-term effects of a single high-intensity exposure to WTC-PM. After 1 month, WTC-PM-exposed wild-type (WT) mice had airway hyperreactivity, whereas RAGE-deficient (Ager-/-) mice were protected. PM-exposed WT mice also had histologic evidence of airspace disease, whereas Ager-/- mice remained unchanged. Inflammatory mediators such as G-CSF (granulocyte colony-stimulating factor), IP-10 (IFN-γ-induced protein 10), and KC (keratinocyte chemoattractant) were differentially expressed after WTC-PM exposure. WTC-PM induced α-SMA, DIAPH1 (protein diaphanous homolog 1), RAGE, and significant lung collagen deposition in WT compared with Ager-/- mice. Compared with WT mice with PM exposure, relative expression of phosphorylated to total CREB (cAMP response element-binding protein) and JNK (c-Jun N-terminal kinase) was significantly increased in the lung of PM-exposed Ager-/- mice, whereas Akt (protein kinase B) was decreased. Random forests of the refined lung metabolomic profile classified subjects with 92% accuracy; principal component analysis captured 86.7% of the variance in three components and demonstrated prominent subpathway involvement, including known mediators of lung disease such as vitamin B6 metabolites, sphingolipids, fatty acids, and phosphatidylcholines. Treatment with a partial RAGE antagonist, pioglitazone, yielded similar fold-change expression of metabolites (N6-carboxymethyllysine, 1-methylnicotinamide, N1+N8-acetylspermidine, and succinylcarnitine [C4-DC]) between WT and Ager-/- mice exposed to WTC-PM. RAGE can mediate WTC-PM-induced airway hyperreactivity and warrants further investigation.
Product Description
Millipore
MILLIPLEX® Mouse Immunoglobulin Isotyping Panel, Mouse IgE Singleplex Kit