MilliporeSigma
  • Simultaneous high-performance liquid chromatographic determination of Cedrus deodara active constituents and their pharmacokinetic profile in mice.
Simultaneous high-performance liquid chromatographic determination of Cedrus deodara active constituents and their pharmacokinetic profile in mice.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-01-15)
B S Sachin, M Koul, A Zutshi, S K Singh, A K Tikoo, M K Tikoo, A K Saxena, S C Sharma, R K Johri
ABSTRACT
A specific and sensitive high-performance liquid chromatographic (HPLC) method with photodiode-array (PDA) ultraviolet detection was developed for the simultaneous determination of three bioactive constituents of Cedrus deodara namely wikstromol, matairesinol and dibenzylbutyrolactol in mouse plasma. In solid-phase extraction (SPE) these constituents were successfully separated using a C18 column by isocratic elution using acetonitrile:water containing hexanesulphonic acid, 32:68 (v/v). The flow rate was set at 1ml/min and detector wavelength at 225nm. Good linearity (r2>0.999) was observed over the studied range of 0.015-5.0microg/ml for wikstromol and 0.030-5.0microg/ml for matairesinol and dibenzylbutyrolactol. The CV values of intra-day precision for wikstromol, matairesinol and dibenzylbutyrolactol were in between 1.8-6.9, 1.7-4.9 and 1.6-4.2% and values of inter-day precision were in between 10.4-12.2, 9.7-11 and 10-11.2%, respectively. The extraction recoveries at low to high concentration were greater than 98, 83 and 87% for each analyte, respectively. The LOQ for wikstromol was 0.015microg/ml and for both matairesinol and dibenzylbutyrolactol it was 0.030microg/ml. The developed method was used to determine the pharmacokinetics of the three analytes in mice after intraperitoneal administration of CD-3.