MilliporeSigma
  • Direct detection of ligand binding to Sepharose-immobilised protein using saturation transfer double difference (STDD) NMR spectroscopy.
Direct detection of ligand binding to Sepharose-immobilised protein using saturation transfer double difference (STDD) NMR spectroscopy.
Biochemical and biophysical research communications (2007-06-19)
Thomas Haselhorst, Anja K Münster-Kühnel, Melanie Oschlies, Joe Tiralongo, Rita Gerardy-Schahn, Mark von Itzstein
ABSTRACT
We report an easy and direct application of 'Saturation Transfer Double Difference' (STDD) NMR spectroscopy to identify ligands that bind to a Sepharose-immobilised target protein. The model protein, cytidine 5'-monophosphate sialic acid (CMP-Sia) synthetase, was expressed as a Strep-Tag II fusion protein and immobilised on Strep-Tactin Sepharose. STD NMR experiments of the protein-enriched Sepharose matrix in the presence of a binding ligand (cytidine 5'-triphosphate, CTP) and a non-binding ligand (alpha/beta-glucose) clearly show that CTP binds to the immobilised enzyme, whereas glucose has no affinity. This approach has three major advantages: (a) only low quantities of protein are required, (b) no specialised NMR technology or the application of additional data analysis by non-routine methods is required, and (c) easy multiple use of the immobilised protein is available.
Sigma-Aldrich
CMP-Sialic Acid Synthetase from Neisseria meningitidis group B, recombinant, expressed in E. coli BL21, ≥10 units/mg protein
Pricing and availability is not currently available.