MilliporeSigma
  • Nonpeptide tachykinin receptor antagonists. II. Pharmacological and pharmacokinetic profile of SB-222200, a central nervous system penetrant, potent and selective NK-3 receptor antagonist.
Nonpeptide tachykinin receptor antagonists. II. Pharmacological and pharmacokinetic profile of SB-222200, a central nervous system penetrant, potent and selective NK-3 receptor antagonist.
The Journal of pharmacology and experimental therapeutics (2000-09-19)
H M Sarau, D E Griswold, B Bush, W Potts, P Sandhu, D Lundberg, J J Foley, D B Schmidt, E F Webb, L D Martin, J J Legos, R G Whitmore, F C Barone, A D Medhurst, M A Luttmann, G A Giardina, D W Hay
ABSTRACT
The pharmacological and pharmacokinetic profile of SB-222200 [(S)-(-)-N-(alpha-ethylbenzyl)-3-methyl-2-phenylquinoline-4-car boxami de], a human NK-3 receptor (hNK-3R) antagonist, was determined. SB-222200 inhibited (125)I-[MePhe(7)]neurokinin B (NKB) binding to Chinese hamster ovary (CHO) cell membranes stably expressing the hNK-3 receptor (CHO-hNK-3R) with a K(i) = 4.4 nM and antagonized NKB-induced Ca(2+) mobilization in HEK 293 cells stably expressing the hNK-3 receptor (HEK 293-hNK-3R) with an IC(50) = 18.4 nM. SB-222200 was selective for hNK-3 receptors compared with hNK-1 (K(i) > 100,000 nM) and hNK-2 receptors (K(i) = 250 nM). In HEK 293 cells transiently expressing murine NK-3 receptors (HEK 293-mNK-3R), SB-222200 inhibited binding of (125)I-[MePhe(7)]NKB (K(i) = 174 nM) and antagonized NKB (1 nM)-induced calcium mobilization (IC(50) = 265 nM). In mice oral administration of SB-222200 produced dose-dependent inhibition of behavioral responses induced by i.p. or intracerebral ventricular administration of the NK-3 receptor-selective agonist, senktide, with ED(50) values of approximately 5 mg/kg. SB-222200 effectively crossed the blood-brain barrier in the mouse and rat. The inhibitory effect of SB-222200 against senktide-induced behavioral responses in the mouse correlated significantly with brain, but not plasma, concentrations of the compound. Pharmacokinetic evaluation of SB-222200 in rat after oral administration (8 mg/kg) indicated sustained plasma concentrations (C(max) = about 400 ng/ml) and bioavailability of 46%. The preclinical profile of SB-222200, demonstrating high affinity, selectivity, reversibility, oral activity, and central nervous system penetration, suggests that it will be a useful tool compound to define the physiological and pathophysiological roles of NK-3 receptors, in particular in the central nervous system.
Sigma-Aldrich
SB 222200, ≥98% (HPLC), solid
Pricing and availability is not currently available.