MilliporeSigma
EXAFS study of zinc coordination in bacitracin A.
Biochimica et biophysica acta (1999-06-03)
F Drabløs, D G Nicholson, M Rønning
ABSTRACT
Bacitracin is a dodecapeptide antibiotic produced by Bacillus sp. The antibacterial activity depends upon the peptide binding a divalent metal. Hitherto, the exact coordination of the cation has not been established. In particular the role played by the sulphur and nitrogen atoms of the thiazoline ring of bacitracin A has not been clear. Here the coordination of Zn2+ by bacitracin A has been studied using extended X-ray absorption fine structure. The experimental data are consistent with a model in which zinc is coordinated by one oxygen and three nitrogen atoms with the sulphur atom of the thiazoline ring not being directly involved in the zinc coordination.
Product Description
Supelco
Bacitracin A, VETRANAL®, analytical standard, main stereoisomer ~50 %