MilliporeSigma
  • Formation of unexpected substitution patterns in sulfonylbutylation of cyclomaltoheptaose promoted by host-guest interaction.
Formation of unexpected substitution patterns in sulfonylbutylation of cyclomaltoheptaose promoted by host-guest interaction.
Carbohydrate research (2000-08-17)
N Rogmann, J Seidel, P Mischnick
ABSTRACT
The distribution of substituents in sulfonylbutylethers of cyclomaltoheptaose (beta-cyclodextrin) formed in aqueous medium has been determined by gas chromatography after hydrolysis and formation of the permethylated sulfonylfluoride derivatives. In contrast to other etherification reactions of beta-cyclodextrin, preferred substitution in position 3 of the glucose units has been detected. From 1H NMR and microcalorimetric experiments, the formation of host-guest complexes by beta-cyclodextrin and the reagent 1,4-butane sultone in water became evident. This spatial preorganization presumably favors the reaction with the O-3. In contrast, in methyl sulfoxide preferred 2-O-alkylation was obtained, indicating that host-guest interaction does not influence regioselectivity in this solvent.
Sigma-Aldrich
1,4-Butane sultone, ≥99%