MilliporeSigma
  • Formation of furan and methylfuran from ascorbic acid in model systems and food.
Formation of furan and methylfuran from ascorbic acid in model systems and food.
Food additives and contaminants (2007-09-22)
A Limacher, J Kerler, B Conde-Petit, I Blank
ABSTRACT
Previous model studies have suggested ascorbic acid as one of the major sources of furan, a possibly hazardous compound found in thermally processed foods (e.g. canned products, jars). The study showed that about 2 mmol mol(-1) furan was obtained when dry-heating ascorbic acid, while much lower amounts were formed upon pressure cooking, i.e. 58 micromol mol(-1) at pH 4 and 3.7 micromol mol(-1) at pH 7. Model reactions also generated 2-methylfuran (MF). However, the MF levels were generally very low with the exception of the binary mixture ascorbic acid/phenylalanine (1 mmol mol(-1)). Studies with 13C-labelled ascorbic acid indicated that furan comprises an intact C4 unit, mainly C-3 to C-6, generated by splitting off two C1 units, i.e. CO2 and formic acid. Possible intermediates are 2-deoxyaldoteroses, 2-furoic acid and 2-furaldehyde, which are known as ascorbic acid degradation products. The mechanism of furan formation from ascorbic acid was validated based on the labelling pattern of furan and the identification of 13CO2 and H13COOH. Furan formation is significantly slowed down in binary mixtures, e.g. the presence of erythrose led to 80% less furan under roasting conditions. This is most likely due to competing reactions in complex systems, thus disfavouring furan formation. The mitigation effect is because furan, contrary to MF, is formed without recombination of ascorbic acid fragments. Therefore, furan levels are definitely much lower in foods than expected from trials with pure ascorbic acid. Consequently, conclusions should be drawn with much caution from model reactions, avoiding extrapolation from oversimplified model systems to food products.
Sigma-Aldrich
2-Methylfuran, contains 200-400 BHT as stabilizer, 99%