MilliporeSigma
Less is more for leaderless mRNA translation.
Molecular cell (2009-02-04)
Daniel N Wilson
ABSTRACT
In this issue of Molecular Cell, Kaberdina et al. (2009) show that prolonged exposure of ribosomes to the antibiotic kasugamycin triggers the spontaneous loss of small subunit proteins and produces a reduced ribosomal particle that exclusively translates leaderless mRNAs.
Sigma-Aldrich
Kasugamycin hydrochloride from Streptomyces kasugaensis