MilliporeSigma
  • Relative contribution of individual oxidized components in ox-LDL to inhibition on endothelium-dependent relaxation in rat aorta.
Relative contribution of individual oxidized components in ox-LDL to inhibition on endothelium-dependent relaxation in rat aorta.
Nutrition, metabolism, and cardiovascular diseases : NMCD (2009-12-17)
W T Wong, C H Ng, S Y Tsang, Y Huang, Z-Y Chen
ABSTRACT
Oxidized low-density lipoprotein (ox-LDL) causes atherosclerosis and endothelial dysfunction. No study up to the present date has examined the relative contribution of all the oxidized components in ox-LDL to inhibition on vascular function. Our aim was to investigate the effects of individual oxidized components at concentrations similar to those in ox-LDL on the impairment of endothelium-dependent relaxation in rat aorta. Rat thoracic aorta was pre-treated with lysophosphatidylcholine (LPC), cholesterol oxidized products (COPs), oxidized linoleic acid (ox-18:2) and oxidized linolenic acid (ox-18:3) at concentrations similar to those in human ox-LDL. Ox-LDL as a whole caused 61% inhibition while LPC, COPs and ox-18:2 at concentrations similar to those in ox-LDL caused 12%, 24% and 19% inhibition, respectively, on endothelium-dependent relaxation, suggesting that COPs produced the most adverse effect followed by ox-18:2 and LPC in an additional way. Three COPs including 7-ketocholesterol, 7α-hydroxycholesterol and 7β-hydroxycholesterol showed inhibition on endothelium-dependent relaxation with E(max) being reduced to 79-87% compared with the control E(max) (95%). At Western blot analysis phosphorylation of eNOS at Ser1177 site and total eNOS were not altered by ox-LDL treatment, indicating that ox-LDL did not affect nitric oxide (NO) synthesis capacity. Ox-LDL might react directly with NO and lower NO bioavailability. The present study demonstrated the relative contribution of individual oxidized components in ox-LDL in the inhibition of endothelium-dependent relaxation in rat aorta. This inhibitory effect could be caused by the reduction of NO bioactivity.
Sigma-Aldrich
7β-Hydroxycholesterol, ≥95%