- Visual detection of dopamine and monitoring tyrosinase activity using a pyrocatechol violet-Sn4+ complex.
Visual detection of dopamine and monitoring tyrosinase activity using a pyrocatechol violet-Sn4+ complex.
Chemical communications (Cambridge, England) (2011-10-25)
Qingling Xu, Juyoung Yoon
PMID22019927
ABSTRACT
In this study, the ensemble of pyrocatechol violet and Sn(4+) was used for dopamine detection in a neutral aqueous solution with high selectivity and sensitivity. The detection course showed three color changes, which could be observed by the naked eye. We also demonstrated that this ensemble can be used to monitor tyrosinase activity.