MilliporeSigma
  • Zinc triflate-promoted glycosidation: synthesis of lipid A disaccharide intermediates.
Zinc triflate-promoted glycosidation: synthesis of lipid A disaccharide intermediates.
Chemical & pharmaceutical bulletin (1992-11-01)
K Nakayama, K Higashi, T Soga, K Uoto, T Kusama
ABSTRACT
Zinc triflate was found to be superior to the heavy metal salts as a promoter in the Koenigs-Knorr type glycosidation reaction in the synthesis of lipid A disaccharide intermediates. It readily promoted the reaction of a complex glycosyl bromide with a reducing sugar moiety and gave the disaccharide with beta-selectivity in good yield. This method would be suitable for the bulk preparation of lipid A disaccharide intermediates.