MilliporeSigma
  • [Effect of catalpol and puerarin freeze-dried powder on coagulability, hemorheology and no in rats with Qi-deficiency and blood-stasis syndrome].
[Effect of catalpol and puerarin freeze-dried powder on coagulability, hemorheology and no in rats with Qi-deficiency and blood-stasis syndrome].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-08-07)
Lijuan Deng, Qin Wang, Huanhuan Yuan, Jialan Liu, Qin Tang, Xiaoyu Xu
ABSTRACT
To study the effect of catalpol and puerarin freeze-dried powder for injection (CPFPI), a new compound traditional Chinese medicine (TCM) preparation, on coagulability, hemorheology and NO in rats with qi-deficiency and blood-stasis syndrome. The model of rats with qi-deficiency and blood-stasis syndrome was established by hunger, fatigue, cold-dampness, panic and high fat diet. Coagulation time (CT) was observed by the glass method, and bleeding time (BT) was measured by tail-cutting method. The effects of CPFPI were also evaluated with prothrombin time (PT), activated partial thromboplastin time (APTT) and thrombin time (TT). HCT was measured by the electric tesistance method, hemorheology indicators were observed by auto-hemorheological instrument. The level of NO in blood serum was measured by NO assay kit. CPFPI 65.40 mg x kg(-1) significantly prolonged CT, BT, PT, APTT and TT in rats. The viscosity of whole blood and plasma, hematocrit, erythrocyte aggregation and rigidity index, and reduced viscosity of whole blood in 65.40 mg x kg(-1) groups were lower than model group. CPFPI 65.40 mg x kg(-1) can raise the level of NO in blood serum. 32.70 mg x kg(-1) markedly prolonged CT, PT and APTT and decreased whole blood viscosity, erythrocyte aggregation index and whole blood reduction viscosity. CPFPI has a significant effect in improving coagulability and hemorheology index and enhancing NO content in blood serum.
Sigma-Aldrich
Catalpol, ≥96% (HPLC)
Pricing and availability is not currently available.