MilliporeSigma
  • Optimization of carbon source for hairy root growth and withaferin A and withanone production in Withania somnifera.
Optimization of carbon source for hairy root growth and withaferin A and withanone production in Withania somnifera.
Natural product communications (2012-11-20)
Ganeshan Sivanandhan, Manoharan Rajesh, Muthukrishnan Arun, Murugaraj Jeyaraj, Gnanajothi Kapil Dev, Markandan Manickavasagam, Natesan Selvaraj, Andy Ganapathi
ABSTRACT
This study optimized carbon sources in half MS liquid medium for maximum biomass accumulation and withanolides production in hairy root culture of Withania somnifera. The highest production of withaferin A and withanone was achieved when sucrose and sucrose+glucose were used individually as carbon sources. The hairy root suspension culture supplemented with a lower level of sucrose (2%) favored hairy root biomass accumulation (1.41 g DW) followed by sucrose+glucose (2+1) when compared with other carbon sources in half MS liquid medium after 40 days of culture. The hairy roots grown on sucrose (4%) enriched half MS liquid medium stimulated higher production of withaferin A (2.21 mg/g DW) and withanone (2.41 mg/g DW) on the 40th day of culture, followed by sucrose+glucose (4+1%) compared with glucose, fructose, maltose and other combinations tested.
Sigma-Aldrich
Withaferin A, ≥95% (HPLC)
Supelco
Withaferin A, analytical standard