MilliporeSigma
  • Solvent and rotational relaxation of coumarin-153 and coumarin-480 in ionic liquid (1-butyl-3-methylimidazolium tetrafluoroborate) modified sodium 1,4-bis(2-ethylhexyl) sulfosuccinate (NaAOT) micelle.
Solvent and rotational relaxation of coumarin-153 and coumarin-480 in ionic liquid (1-butyl-3-methylimidazolium tetrafluoroborate) modified sodium 1,4-bis(2-ethylhexyl) sulfosuccinate (NaAOT) micelle.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-12)
Vishal Govind Rao, Chiranjib Banerjee, Sarthak Mandal, Surajit Ghosh, Nilmoni Sarkar
ABSTRACT
Understanding ion transport dynamics, structure of surfactant aggregates in ionic liquids or ionic liquid/water solutions are quite interesting and potentially important due to widespread applications of surfactant-based systems. In this manuscript we have investigated the effect of 1-butyl-3-methylimidazolium tetrafluoroborate (bmimBF(4)) addition on solvent and rotational relaxation of coumarin-153 (C-153) and coumarin-480 (C-480) in aqueous solution of sodium 1,4-bis(2-ethylhexyl) sulfosuccinate (NaAOT) using steady state and picosecond time resolved fluorescence spectroscopy. The strong adsorption of the bmim(+) at the interface and the role of the ionic liquid particularly the cation bmim(+) in the modification of the interfacial geometry were probed by the analysis of decay parameters and the rotational relaxation parameters. Since the addition of the NaAOT in water-bmimBF(4) mixture above critical micellar concentration (48 mM, obtained from observing pyrene fluorescence) causes strong adsorption of the ionic liquid particularly the cation bmim(+), the average solvation time, particularly the slow component increases significantly. More importantly we have found the probe dependent solvation dynamics due to the different location of the probe molecules, C-153 and C-480. C-153 being hydrophobic in nature resides in the stern layer and the adsorption of the bmim(+) at the interface modifies stern layer more effectively. So we have observed more pronounced change in solvation dynamics in case of C-153 compared to that in case of C-480. The fluorescence anisotropy decays of the probe molecules were found to be biexponential in nature. The anisotropy decay was interpreted by using a model which consists of the wobbling (rotational) and translational diffusion of the dye coupled with the rotational motion of the micelle as a whole.
Sigma-Aldrich
Sulfosuccinic acid solution, 70 wt. % in H2O
Pricing and availability is not currently available.