MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Biocompatibility of boron nitride nanotubes: an up-date of in vivo toxicological investigation.
Biocompatibility of boron nitride nanotubes: an up-date of in vivo toxicological investigation.
International journal of pharmaceutics (2013-01-30)
Gianni Ciofani, Serena Danti, Simone Nitti, Barbara Mazzolai, Virgilio Mattoli, Mario Giorgi
ABSTRACT
Boron nitride nanotubes (BNNTs) represent an innovative and extremely intriguing class of nanomaterials, and preliminar but encouraging studies about their applications in biomedicine have emerged in the latest years. As a consequence, a systematic investigation of their biosafety has become of fundamental importance in the biomedical research. Extending our previous pilot in vivo study, here we present biocompatibility data of BNNTs injected in rabbits at a dose up to 10mg/kg. No significant adverse effects were found up to 7 days since their administration, and no impairments in blood, liver and kidney functionality were highlighted. Moreover, a terminal half-life circulation of about 90min was found. All the collected data are very promising, suggesting the optimal biocompatibility of BNNTs, and thus enabling their exploitation in nanomedicine as nanotransducers and nanocarriers.
Sigma-Aldrich
Boron nitride, powder, ~1 μm, 98%