MilliporeSigma
  • [Excretory insufficiency of the pancreas after occlusion of the ductal system with glue].
[Excretory insufficiency of the pancreas after occlusion of the ductal system with glue].
Vestnik khirurgii imeni I. I. Grekova (1987-03-01)
V S Zemskov, V S Shchitov, V I Biletskiĭ, S N Panchenko, V N Blagodarov
Sigma-Aldrich
Poly(propylene glycol), tolylene 2,4-diisocyanate terminated, average Mn ~2,300 (narrow MW distribution), isocyanate ~3.6 wt. %