MilliporeSigma
  • Suggested minimal effective dose of risperidone based on PET-measured D2 and 5-HT2A receptor occupancy in schizophrenic patients.
Suggested minimal effective dose of risperidone based on PET-measured D2 and 5-HT2A receptor occupancy in schizophrenic patients.
The American journal of psychiatry (1999-06-09)
S Nyberg, B Eriksson, G Oxenstierna, C Halldin, L Farde
ABSTRACT
Multicenter trials with the novel antipsychotic risperidone have suggested a standard dose of 6 mg/day. However, a dose producing the highest response rate in fixed-dose studies is likely to exceed the minimal effective dose in most patients. The aim of this positron emission tomography (PET) study was to suggest a minimal effective dose of risperidone based on measurements of dopamine D2 and serotonin 5-HT2A receptor occupancy. Eight first-episode or drug-free schizophrenic patients were treated with risperidone, 6 mg/day, for 4 weeks and then 3 mg/day for 2 weeks. PET was performed after 4 and 6 weeks, with [11C]raclopride to measure D2 receptor occupancy and [11C]N-methylspiperone to measure 5-HT2A receptor occupancy. Seven patients completed the study and responded to treatment with risperidone. No patient had extrapyramidal side effects at the time of inclusion in the study. At the 6-mg/day dose, mean D2 receptor occupancy was 82% (range = 79%-85%), 5-HT2A receptor occupancy was 95% (range = 86%-109%), and six patients had developed extrapyramidal side effects. After dose reduction to 3 mg/day, D2 receptor occupancy was 72% (range = 53%-78%), and 5-HT2A receptor occupancy was 83% (range = 65%-112%). Three patients had extrapyramidal side effects at this time. Treatment with risperidone, 6 mg/day, is likely to induce unnecessarily high D2 receptor occupancy, with a consequent risk of extrapyramidal side effects. High 5-HT2A receptor occupancy did not prevent extrapyramidal side effects completely. The authors previously suggested an optimal interval for D2 receptor occupancy of 70%-80%. To achieve this, resperidone, 4 mg/day, should be a suitable initial dose for antipsychotic effect with a minimal risk of extrapyramidal side effects in most patients.