MilliporeSigma
  • Biphenylacetic acid enhances the antagonistic action of fluoroquinolones on the GABA(A)-mediated responses of the isolated guinea-pig ileum.
Biphenylacetic acid enhances the antagonistic action of fluoroquinolones on the GABA(A)-mediated responses of the isolated guinea-pig ileum.
Pharmacological research (2001-09-01)
M Koutsoviti-Papadopoulou, E Nikolaidis, G Kounenis
ABSTRACT
This paper examines the effect of biphenylacetic acid on the antagonistic action of norfloxacin and enoxacin on the GABA(A)-mediated responses of the isolated guinea-pig ileum. GABA produced transient contractions followed by relaxation. The contractile effect of exogenously applied GABA was concentration-dependent with EC(50)= 9.8 x 10(-6) M. This contractile effect was not significantly modified by biphenylacetic acid, and the EC(50) value for GABA in the presence of 10(-5) M biphenylacetic acid was 1.15 x 10(-5) M. The GABA contractile effect was inhibited, dose-dependently, by either norfloxacin or enoxacin, but only at concentrations higher than 10(-5) M. The response of the ileum to GABA (at EC(50)) was reduced to 35 and 36% by pretreatment with 10(-5) M norfloxacin or enoxacin, respectively. However, in the presence of 10(-5) M biphenylacetic acid, the response of the ileum to GABA was reduced to 2.2% by pretreatment with 10(-5) M enoxacin, while it was completely abolished by pretreatment with 10(-5) M norfloxacin and the IC(50) values were 5.5 x 10(-7) and 1.5 x 10(-6) M for norfloxacin and enoxacin, respectively. These data show that biphenylacetic acid whilst having no effect at the GABA(A)-mediated contractile response of the guinea-pig ileum, enhances the antagonistic effect of both enoxacin and norfloxacin. This suggests that combined administration of fluoroquinolones and biphenylacetic acid synergistically inhibits GABA(A)-receptors at the intestinal level.
Sigma-Aldrich
4-Biphenylacetic acid, 98%