MilliporeSigma
  • Human serial learning: enhancement with arecholine and choline impairment with scopolamine.
Human serial learning: enhancement with arecholine and choline impairment with scopolamine.
Science (New York, N.Y.) (1978-07-21)
N Sitaram, H Weingartner, J C Gillin
PMID351808
ABSTRACT
Arecholine (4 milligrams), a cholinergic agonist, and choline (10 grams), a precursor of acetylcholine, significantly enhanced serial learning in normal human subjects. The subjects received methscopolamine prior to both arecholine and placebo injections. Conversely, scopolamine (0.5 milligram), a cholinergic antagonist, impaired learning and this impairment was reversed by arecholine and choline and the impairment after scopolamine were inversely proportional to the subject's performance on placebo; that is, "poor" performers were more vulnerable to both the enhancing effect of cholinergic agonist and precursor and the impairment after cholinergic antagonist than "good" performers.