MilliporeSigma
  • Critical evaluation of micellization behavior of nonionic surfactant MEGA 10 in comparison with ionic surfactant tetradecyltriphenylphosphonium bromide studied by microcalorimetric method in aqueous medium.
Critical evaluation of micellization behavior of nonionic surfactant MEGA 10 in comparison with ionic surfactant tetradecyltriphenylphosphonium bromide studied by microcalorimetric method in aqueous medium.
The journal of physical chemistry. B (2006-05-19)
Madhumita Prasad, Indranil Chakraborty, Animesh K Rakshit, Satya P Moulik
ABSTRACT
The micellization behavior of MEGA 10 has been studied at nine different temperatures by isothermal titration calorimetry (ITC), and thermodynamics of the process have been evaluated and examined in detail. The aggregation number of the nonionic surfactant has been estimated from the ITC results by a simulation procedure based on the mass action principle of micellization of the surfactant. The cmc of MEGA 10 has shown a minimum in temperature dependence as observed for ionic surfactants. For a comparison, the cmc and related thermodynamic parameters of an ionic surfactant, tetradecyltriphenylphosphonium bromide (C(14)TPB) studied at several temperatures in aqueous medium has been considered. The contributions of the headgroups of both the surfactants to the free energies of their respective micellization have been deciphered and presented.
Sigma-Aldrich
N-Decanoyl-N-methylglucamine, ≥98% (GC)