MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Access to enantiomerically enriched cis-2,3-disubstituted azetidines via diastereoselective hydrozirconation.
Access to enantiomerically enriched cis-2,3-disubstituted azetidines via diastereoselective hydrozirconation.
Organic letters (2011-03-10)
Tarun K Pradhan, K Syam Krishnan, Jean-Luc Vasse, Jan Szymoniak
ABSTRACT
An asymmetric variant of the hydrozirconation reaction has been established starting from Boc-protected chiral allylic amines. The resulting diastereoselectively formed N-functionalized organozirconiums can be considered as promising chirons. In this case, they have been transformed into enantiomerically enriched cis-2,3-disubstituted azetidines through a iodination/cyclization sequence.