MilliporeSigma
New solid phase synthesis of distamycin analogues.
Molecules (Basel, Switzerland) (2011-04-13)
Danuta Drozdowska, Drozdowska Danuta
ABSTRACT
A novel and straightforward solid phase synthesis of distamycin analogues containing benzene units has been developed.