MilliporeSigma
  • Heterochromatin patterns and ribosomal DNA loci distribution in diploid and polyploid Crotalaria species (Leguminosae, Papilionoideae), and inferences on karyotype evolution.
Heterochromatin patterns and ribosomal DNA loci distribution in diploid and polyploid Crotalaria species (Leguminosae, Papilionoideae), and inferences on karyotype evolution.
Genome (2011-08-26)
Mateus Mondin, Margarida L R Aguiar-Perecin
ABSTRACT
Most Crotalaria species display a symmetric karyotype with 2n = 16, but 2n = 14 is found in Chrysocalycinae subsection Incanae and 2n = 32 in American species of the section Calycinae. Seven species of the sections Chrysocalycinae, Calycinae, and Crotalaria were analyzed for the identification of heterochromatin types with GC- and AT-specific fluorochromes and chromosomal location of ribosomal DNA loci using fluorescent in situ hybridization (FISH). A major 45S rDNA locus was observed on chromosome 1 in all the species, and a variable number of minor ones were revealed. Only one 5S rDNA locus was observed in the species investigated. Chromomycin A(3) (CMA) revealed CMA(+) bands colocalized with most rDNA loci, small bands unrelated to ribosomal DNA on two chromosome pairs in Crotalaria incana, and CMA(+) centromeric bands that were quenched by distamycin A were detected in species of Calycinae and Crotalaria sections. DAPI(+) bands were detected in C. incana. The results support the species relationships based on flower specialization and were useful for providing insight into mechanisms of karyotype evolution. The heterochromatin types revealed by fluorochromes suggest the occurrence of rearrangements in repetitive DNA families in these heterochromatic blocks during species diversification. This DNA sequence turnover and the variability in number/position of rDNA sites could be interpreted as resulting from unequal crossing over and (or) transposition events. The occurrence of only one 5S rDNA locus and the smaller chromosome size in the polyploids suggest that DNA sequence losses took place following polyploidization events.