MilliporeSigma
  • The JAZ proteins: a crucial interface in the jasmonate signaling cascade.
The JAZ proteins: a crucial interface in the jasmonate signaling cascade.
The Plant cell (2011-10-04)
Laurens Pauwels, Alain Goossens
ABSTRACT
Jasmonates are phytohormones that regulate many aspects of plant growth, development, and defense. Within the signaling cascades that are triggered by jasmonates, the JASMONATE-ZIM DOMAIN (JAZ) repressor proteins play a central role. The endogenous bioactive JA-Ile conjugate mediates the binding of JAZ proteins to the F-box protein CORONATINE INSENSITIVE1 (COI1), part of the Skp1/Cullin/F-box SCF(COI1) ubiquitin E3 ligase complex. Upon the subsequent destruction of the JAZ proteins by the 26S proteasome, multiple transcription factors are relieved from JAZ-mediated repression, allowing them to activate their respective downstream responses. However, many questions remain regarding the targets, specificity, function, and regulation of the different JAZ proteins. Here, we review recent studies on the model plant Arabidopsis thaliana that provided essential and novel insights. JAZ proteins have been demonstrated to interact with a broad array of transcription factors that each control specific downstream processes. Recruitment of the corepressor TOPLESS unveiled a mechanism for JAZ-mediated gene repression. Finally, the presence of JAZ proteins was also found to be regulated by alternative splicing and interactions with proteins from other hormonal signaling pathways. Overall, these contemporary findings underscore the value of protein-protein interaction studies to acquire fundamental insight into molecular signaling pathways.