MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • pH-dependent, thermosensitive polymeric nanocarriers for drug delivery to solid tumors.
pH-dependent, thermosensitive polymeric nanocarriers for drug delivery to solid tumors.
Biomaterials (2013-03-19)
Ching-Yi Chen, Tae Hee Kim, Wen-Chung Wu, Chi-Ming Huang, Hua Wei, Christopher W Mount, Yanqing Tian, Sei-Hum Jang, Suzie H Pun, Alex K-Y Jen
ABSTRACT
Polymeric micelles are promising carriers for anti-cancer agents due to their small size, ease of assembly, and versatility for functionalization. A current challenge in the use of polymeric micelles is the sensitive balance that must be achieved between stability during prolonged blood circulation and release of active drug at the tumor site. Stimuli-responsive materials provide a mechanism for triggered drug release in the acidic tumor and intracellular microenvironments. In this work, we synthesized a series of dual pH- and temperature-responsive block copolymers containing a poly(ε-caprolactone) (PCL) hydrophobic block with a poly(triethylene glycol) block that were copolymerized with an amino acid-functionalized monomer. The block copolymers formed micellar structures in aqueous solutions. An optimized polymer that was functionalized with 6-aminocaproic acid (ACA) possessed pH-sensitive phase transitions at mildly acidic pH and body temperature. Doxorubicin-loaded micelles formed from these polymers were stable at blood pH (~7.4) and showed increased drug release at acidic pH. In addition, these micelles displayed more potent anti-cancer activity than free doxorubicin when tested in a tumor xenograft model in mice.
Product Description
Supelco
Aminocaproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Polycaprolactone, average Mn 45,000
Sigma-Aldrich
6-Aminocaproic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Sigma-Aldrich
6-Aminocaproic acid, BioUltra, ≥99%
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)
Sigma-Aldrich
Polycaprolactone, average Mw ~14,000, average Mn ~10,000 by GPC
Sigma-Aldrich
Polycaprolactone, average Mn 80,000
Sigma-Aldrich
Triethylene glycol, BioUltra, anhydrous, ≥99.0% (GC)
Sigma-Aldrich
Triethylene glycol, ReagentPlus®, 99%