MilliporeSigma
  • The cycle characteristics of clomiphene with clomiphene and menotropins in polycystic ovary syndrome and non polycystic ovary syndrome infertile patients.
The cycle characteristics of clomiphene with clomiphene and menotropins in polycystic ovary syndrome and non polycystic ovary syndrome infertile patients.
Minerva ginecologica (2013-05-22)
M Ghasemi, H Ashraf, H Koushyar, N Mousavifar
ABSTRACT
This study compares the cycle characteristics of clomiphene (CC) with CC+HMG (Human Menopausal Gonadotropin or Menotropins) in Polycystic Ovary Syndrome (PCOS) and non-PCOS infertile patients. Patients were treated by CC + minimal HMg protocol. The cancellation rate, the mean number of different follicle sizes and endometrial thickness and pattern were compared. The cancelled cycles due to non-responsiveness were significantly higher in CC compared to CC+ minimal HMg protocol. PCOS patients are significantly nonresponsive in CC cycle and hyperresponsive in CC+ minimal HMg cycles. The mean number of different sizes of follicles and the endometrial thickness were significantly higher in CC+ minimal HMg. PCOS patients were significantly different from non-PCOS regarding the number of mature follicle and endometrial thickness. The pregnancy rate was 11% (10.2% in non-PCOS and 12.2% in PCOS). CC+ minimal HMg is a viable alternative to HMg /FSH only protocol in CC failure or resistant patients, and its efficacy can be mostly attributed to improvement of endometrial quality and increase in follicle number. Moreover, due to high cancellation of PCOS patients treated by this protocol, seemingly other alternatives should be found; perhaps sequential letrozole+HMg/FSH that have been shown to improve the ovarian response in this group of patients.