MilliporeSigma
  • Physical and biological consequences of incorporation of antiviral agents into virus DNA.
Physical and biological consequences of incorporation of antiviral agents into virus DNA.
Antiviral research (1984-12-01)
W H Prusoff, W R Mancini, T S Lin, J J Lee, S A Siegel, M J Otto
ABSTRACT
The molecular basis for the antiviral activity is discussed for a variety of nucleoside compounds approved for clinical use in the U.S.A. (5-iodo-2'-deoxyuridine, 5-trifluoromethyl-2'-deoxyuridine, 9-beta-D-arabinofuranosyladenine, 9-(2-hydroxyethoxymethyl)guanine), or in clinical trial (E-5-(2-bromovinyl)-2'-deoxyuridine, 1-(2-deoxy-2-fluoro-beta-D-arabinosyl)-5-iodocytosine, 1-beta-D-ribofuranosyl-1,2,4-triazole-3-carboxamide), or of specific interest to our laboratory (5-iodo-5'-amino-2',5'-dideoxyuridine, 5'-amino-5'-deoxythymidine). The consequence of incorporation of idoxuridine, the 5'-amino analog of thymidine or the 5'-amino analog of idoxuridine into the DNA of herpes simplex virus type 1 on transcription and translation is emphasized.