MilliporeSigma
  • In vitro effect of adenosine agonist GR79236 on the insulin sensitivity of glucose utilisation in rat soleus and human rectus abdominus muscle.
In vitro effect of adenosine agonist GR79236 on the insulin sensitivity of glucose utilisation in rat soleus and human rectus abdominus muscle.
Biochimica et biophysica acta (1996-06-07)
J M Webster, L Heseltine, R Taylor
ABSTRACT
The dose-response effects of a new adenosine agonist, GR79236, were examined in isolated rat soleus muscle strips and human rectus abdominus muscle strips. Effects on the insulin sensitivity of carbohydrate metabolism were examined, in particular upon insulin stimulated glycogen synthesis and glycolytic flux. In the presence of adenosine deaminase (ADA), GR79236 increased insulin sensitivity of pyruvate release from rat soleus muscle strips by 24% from 82.5 +/- 10.0 to 102.5 +/- 10.0 (P < 0.01), by 27% to 105.0 +/- 12.5 (P < 0.01) and by 24% to 102.5 +/- 10.0 (P < 0.01) nmol/25 mg per h at 0.1 and 10 microM GR79236, respectively. Rates of lactate release followed a similar but non-significant trend. Addition of GR79236 in the presence of ADA had no effect on rates of glycogen synthesis. Insulin stimulated rates of pyruvate or lactate release or of glycogen synthesis were unaffected by the addition of adenosine deaminase or GR79236 in human rectus abdominus muscle strips. Adenosine agonists may act indirectly to modulate insulin sensitivity of carbohydrate metabolism.