MilliporeSigma
  • Differential regulation of metallothionein genes in rainbow trout fibroblasts, RTG-2.
Differential regulation of metallothionein genes in rainbow trout fibroblasts, RTG-2.
Biochimica et biophysica acta (1990-07-30)
M Zafarullah, P E Olsson, L Gedamu
ABSTRACT
Exposure of a trout gonadal fibroblast (RTG-2) cell line to ZnCl2, CdCl2 and CuCl2 resulted in differential levels of accumulation of metallothionein (MT) mRNA. ZnCl2 being the most effective agent induced MT mRNA in 3 h, with 172-fold induction after 48 h and continued accumulation up to 144 h. Following CdCl2 treatment, mRNA could be detected after 24 h, reaching peak levels at 72 h. Furthermore, trout MT mRNA could be detected up to 8 days after withdrawal of extraneous ZnCl2. Using a novel technique of primer extension and DNA sequencing with total RNA as template, specificity of the trout MTa and MTb gene-specific primers was established. Primer extension studies revealed a higher response of MTa to ZnCl2 and CdCl2 compared to MTb. Insensitivity of MT mRNA induction to cycloheximide suggested that induction by the metals was independent of de novo protein synthesis. However, simultaneous exposure of cells to actinomycin D and metals completely inhibited MT mRNA synthesis implying control at the transcriptional level.
Product Description
RTG-2, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 90102529, fish ovary, Fibroblast