  • Search Results
  • GC-MS analysis of acylated derivatives of the side-chain regioisomers of 4-methoxy-3-methyl-phenethylamines related to methylenedioxymethamphetamine.
GC-MS analysis of acylated derivatives of the side-chain regioisomers of 4-methoxy-3-methyl-phenethylamines related to methylenedioxymethamphetamine.
Journal of chromatographic science (2007-11-21)
Tamer Awad, C Randall Clark, Jack DeRuiter
ABSTRACT
The five side-chain regioisomers of 4-methoxy-3-methylphenethylamine constitute a unique set of compounds having an isobaric relationship with the controlled drug substance 3,4-methylenedioxymethamphetamine (3,4-MDMA or Ecstasy). These isomeric forms of the 4-methoxy-3-methylphenethylamines have mass spectra essentially equivalent to 3,4-MDMA, and all have a molecular weight of 193 and major fragment ions in their electron ionization mass spectra at m/z 58 and 135/136. Mass spectral differentiation of 2,3- and 3,4-MDMA from primary and secondary amine regioisomeric side chains of 4-methoxy-3-methylphenethylamines was possible after formation of the perfluoroacyl derivatives, pentafluoropropionamides and heptafluorobutyrylamides. The mass spectra for these derivatives are significantly individualized, and the resulting unique fragment ions allow for specific side-chain identification. The individualization is the result of fragmentation of the alkyl carbon-nitrogen bond, which yielded unique hydrocarbon fragments. The heptafluorobutyrylamide derivatives offer more fragment ions for molecular individualization among these regioisomeric substances. Gas chromatographic separation on relatively non-polar stationary phases successfully resolves these derivatives.
Sigma-Aldrich
4-Methoxy-3-methylbenzaldehyde, 99%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.