MilliporeSigma
  • Combination corticosteroid/β-agonist inhaler as reliever therapy: a solution for intermittent and mild asthma?
Combination corticosteroid/β-agonist inhaler as reliever therapy: a solution for intermittent and mild asthma?
The Journal of allergy and clinical immunology (2013-12-29)
Richard Beasley, Mark Weatherall, Philippa Shirtcliffe, Robert Hancox, Helen K Reddel
ABSTRACT
The recommended treatment of mild asthma is regular maintenance inhaled corticosteroids (ICSs) with a short-acting β-agonist as a separate inhaler used when needed for symptom relief. However, the benefits of regular ICS use in actual clinical practice are limited by poor adherence and low prescription rates. An alternative strategy would be the symptom-driven (as-required or "prn") use of a combination ICS/short-acting β-agonist or ICS/long-acting β-agonist inhaler as a reliever rather than regular maintenance use. The rationale for this approach is to titrate both the ICS and β-agonist dose according to need and enhance ICS use in otherwise poorly adherent patients who overrely on their reliever β-agonist inhaler. This strategy will only work if the β-agonist component has a rapid onset of action for symptom relief. There is evidence to suggest that this regimen has advantages over regular ICS therapy and might represent an effective, safe, and novel therapy for the treatment of intermittent and mild asthma. In this commentary we review this evidence and propose that randomized controlled trials investigating different combination ICS/β-agonist inhaler products prescribed according to this regimen in intermittent and mild asthma are an important priority.
Product Description
Budesonide, European Pharmacopoeia (EP) Reference Standard
Supelco
Budesonide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Budesonide, ≥99%