MilliporeSigma
  • Separation and Cultivation of Hematopoietic Stem Cells from Umbilical Cord Blood by Permeation through Membranes with Nano-Segments
Separation and Cultivation of Hematopoietic Stem Cells from Umbilical Cord Blood by Permeation through Membranes with Nano-Segments
Current Science (2011)
Higuchi, Akon; Chen, Li-Ying; Shiao, Jui-Shiang; Ling, Qing-Dong; Ko